Product Details

Assay Reference Genome
Location

Chr.3:174464967 on build GRCh38
Cytoband
3q26.31
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
254827
Gene Symbol
NAALADL2
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 2
Gene Aliases
Location
Chr.3:174439669-175809510 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NAALADL2 XM_006713560.2 XP_006713623.1
XM_017006075.1 XP_016861564.1
XM_017006077.1 XP_016861566.1
XM_017006078.1 XP_016861567.1
XM_017006080.1 XP_016861569.1
XM_017006082.1 XP_016861571.1
AJ607398.1
AJ607399.1
AJ607401.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1005730 Chr3:174266156 - 174829055 on Build GRCh38 Gain NLGN1 NAALADL2
nsv4124 Chr3:174461292 - 174478318 on Build GRCh38 Deletion NAALADL2
nsv520908 Chr3:174083961 - 174816144 on Build GRCh38 Loss NLGN1 NAALADL2

View Full Product Details