Product Details

Assay Reference Genome
Location

Chr.22:33165977 on build GRCh38
Cytoband
22q12.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
9215
Gene Symbol
LARGE1
Gene Name
LARGE xylosyl- and glucuronyltransferase 1
Gene Aliases
LARGE, MDC1D, MDDGA6, MDDGB6
Location
Chr.22:33162237-33922841 on build GRCh38
Assay Gene Location
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LARGE1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3893476 Chr22:33163488 - 33370369 on Build GRCh38 Gain LARGE1
esv3568374 Chr22:33152451 - 33186805 on Build GRCh38 Loss LARGE1
nsv1056019 Chr22:33131887 - 33261303 on Build GRCh38 Gain LARGE1

View Full Product Details