Product Details

Assay Reference Genome
Location

Chr.14:77981686 on build GRCh38
Cytoband
14q24.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
9369
Gene Symbol
NRXN3
Gene Name
neurexin 3
Gene Aliases
C14orf60
Location
Chr.14:77979904-79868290 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NRXN3 XM_017021790.1 XP_016877279.1
XM_017021791.1 XP_016877280.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv34345 Chr14:77904112 - 78030244 on Build GRCh38 Loss ADCK1 NRXN3
esv2759999 Chr14:77912813 - 78035676 on Build GRCh38 Loss ADCK1 NRXN3

View Full Product Details