Product Details

Assay Reference Genome
Location

Chr.11:78666506 on build GRCh38
Cytoband
11q14.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
26011
Gene Symbol
TENM4
Gene Name
teneurin transmembrane protein 4
Gene Aliases
Doc4, ETM5, ODZ4, TNM4, Ten-M4
Location
Chr.11:78653283-79441019 on build GRCh38
Assay Gene Location
Within Intron 34
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
TENM4 NM_001098816.2 NP_001092286.2
XM_011544933.2 XP_011543235.1
XM_017017525.1 XP_016873014.1
XM_017017526.1 XP_016873015.1
AB037723.3 BAA92540.3
AK056531.1 BAB71206.1
AK127101.1
AK127705.1
BX537983.1 CAD97943.1
BX640737.1 CAE45850.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv483084 Chr11:78645595 - 78818126 on Build GRCh38 Loss TENM4
nsv555455 Chr11:78629643 - 78671072 on Build GRCh38 Gain TENM4
esv34127 Chr11:78661992 - 79102008 on Build GRCh38 Loss TENM4

View Full Product Details