Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation ID |
Location |
CNV Subtype |
Genes |
nsv830674 |
Chr6:63645507 - 63825908 on Build GRCh38 |
Gain+Loss |
EYS
PHF3
|
Product Details
- Assay Reference Genome
Location
Chr.6:63735106 on build GRCh38
- Cytoband
- 6q12
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 346007
- Gene Symbol
- EYS
- Gene Name
- eyes shut homolog (Drosophila)
- Gene Aliases
- C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
- Location
- Chr.6:63719980-65707225 on build GRCh38
- Assay Gene Location
- Within Intron 44
Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation |
Location |
CNV Subtype |
Genes |
nsv830674 |
Chr6:63645507 - 63825908 on Build GRCh38 |
Gain+Loss |
EYS
PHF3
|
View Full Product Details