Product Details

Assay Reference Genome
Location

Chr.6:63741765 on build GRCh38
Cytoband
6q12
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
346007
Gene Symbol
EYS
Gene Name
eyes shut homolog (Drosophila)
Gene Aliases
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
Location
Chr.6:63719980-65707225 on build GRCh38
Assay Gene Location
Within Intron 44
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
EYS NM_001142800.1 NP_001136272.1
NM_001292009.1 NP_001278938.1
FJ416331.1
FM209056.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv830674 Chr6:63645507 - 63825908 on Build GRCh38 Gain+Loss PHF3 EYS

View Full Product Details