Product Details

Assay Reference Genome
Location

Chr.3:188161125 on build GRCh38
Cytoband
3q27.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
4026
Gene Symbol
LPP
Gene Name
LIM domain containing preferred translocation partner in lipoma
Gene Aliases
Location
Chr.3:188153050-188890671 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LPP NM_001167672.2 NP_001161144.1
XM_005247446.4 XP_005247503.1
XM_005247450.4 XP_005247507.1
XM_005247451.4 XP_005247508.1
XM_011512820.2 XP_011511122.1
XM_011512827.2 XP_011511129.1
XM_011512828.2 XP_011511130.1
XM_011512831.2 XP_011511133.1
XM_011512833.2 XP_011511135.1
XM_011512834.2 XP_011511136.1
XM_017006377.1 XP_016861866.1
XM_017006379.1 XP_016861868.1
XM_017006380.1 XP_016861869.1
AK304035.1
AL833171.1
BX474180.1
CD697098.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv34193 Chr3:188129943 - 188550793 on Build GRCh38 Loss FLJ42393 LPP LPP-AS2
nsv1003486 Chr3:188157145 - 188284087 on Build GRCh38 Gain FLJ42393 LPP
esv2751987 Chr3:188124933 - 188289510 on Build GRCh38 Gain FLJ42393 LPP LPP-AS2

View Full Product Details