Product Details

Assay Reference Genome
Location

Chr.3:59756350 on build GRCh38
Cytoband
3p14.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2272
Gene Symbol
FHIT
Gene Name
fragile histidine triad
Gene Aliases
AP3Aase, FRA3B
Location
Chr.3:59747278-61251474 on build GRCh38
Assay Gene Location
Within Intron 15
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
FHIT NM_001166243.2 NP_001159715.1
NM_001320899.1 NP_001307828.1
NM_001320900.1 NP_001307829.1
NM_001320901.1 NP_001307830.1
NM_002012.3 NP_002003.1
NR_135491.1
XM_017005880.1 XP_016861369.1
XM_017005881.1 XP_016861370.1
XM_017005882.1 XP_016861371.1
XM_017005883.1 XP_016861372.1
XM_017005884.1 XP_016861373.1
XM_017005885.1 XP_016861374.1
XM_017005886.1 XP_016861375.1
AK127931.1
AK289824.1
BC032336.1 AAH32336.1
DB131595.1
DQ120721.1 AAZ23623.1
KJ534835.1
U46922.1 AAA99013.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv4749n100 Chr3:58504269 - 60219035 on Build GRCh38 Gain LOC339902 C3orf67-AS1 C3orf67 FAM3D FAM107A ACOX2 FAM3D-AS1 FHIT
nsv1014601 Chr3:59680883 - 59902440 on Build GRCh38 Loss FHIT

View Full Product Details