Product Details

Assay Reference Genome
Location

Chr.2:51073762 on build GRCh38
Cytoband
2p16.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
730100
Gene Symbol
LOC730100
Gene Name
uncharacterized LOC730100
Gene Aliases
Location
Chr.2:51032601-52407917 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LOC730100 NR_135237.1
AK127244.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3892204 Chr2:51065295 - 51119864 on Build GRCh38 Loss LOC730100
nsv1000086 Chr2:50879876 - 51193400 on Build GRCh38 Loss NRXN1 LOC730100
esv2759048 Chr2:51054576 - 51138744 on Build GRCh38 Gain LOC730100
nsv581868 Chr2:51053379 - 51125305 on Build GRCh38 Loss LOC730100
nsv521850 Chr2:51052256 - 51210278 on Build GRCh38 Loss LOC730100
dgv3814n100 Chr2:51044124 - 51123977 on Build GRCh38 Loss LOC730100
dgv1909n106 Chr2:51070139 - 51156062 on Build GRCh38 Deletion LOC730100
nsv1012208 Chr2:51039660 - 51188856 on Build GRCh38 Loss LOC730100
esv3892237 Chr2:51020156 - 51130764 on Build GRCh38 Loss NRXN1 LOC730100
dgv6866n54 Chr2:50820175 - 51108720 on Build GRCh38 Loss NRXN1 LOC730100
nsv581848 Chr2:50711821 - 51118377 on Build GRCh38 Loss NRXN1 LOC730100
nsv581853 Chr2:50786509 - 51326909 on Build GRCh38 Loss NRXN1 LOC730100
esv2763321 Chr2:50610356 - 51126248 on Build GRCh38 Loss NRXN1 MIR8485 LOC730100
esv3892215 Chr2:50887566 - 51133529 on Build GRCh38 Loss NRXN1 LOC730100
esv3590745 Chr2:51071287 - 51110515 on Build GRCh38 Loss LOC730100
nsv834014 Chr2:50954708 - 51133399 on Build GRCh38 Loss NRXN1 LOC730100
nsv1002086 Chr2:50685023 - 51177923 on Build GRCh38 Loss NRXN1 MIR8485 LOC730100
nsv1000966 Chr2:51071789 - 51108905 on Build GRCh38 Loss LOC730100

View Full Product Details