Product Details

Assay Reference Genome
Location

Chr.5:58971157 on build GRCh38
Cytoband
5q11.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
5144
Gene Symbol
PDE4D
Gene Name
phosphodiesterase 4D
Gene Aliases
ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
Location
Chr.5:58969038-60488098 on build GRCh38
Assay Gene Location
Within Exon 34
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PDE4D NM_001104631.1 15 6109 NP_001098101.1
NM_001165899.1 17 6000 NP_001159371.1
NM_001197218.1 15 5848 NP_001184147.1
NM_001197219.1 15 5626 NP_001184148.1
NM_001197220.1 15 6264 NP_001184149.1
NM_001197221.1 11 5584 NP_001184150.1
NM_001197222.1 10 5670 NP_001184151.1
NM_001197223.1 10 5460 NP_001184152.1
NM_006203.4 15 5652 NP_006194.2
XM_005248537.3 15 6806 XP_005248594.1
XM_005248538.4 16 8707 XP_005248595.1
XM_011543469.1 18 6089 XP_011541771.1
XM_011543470.2 18 6013 XP_011541772.1
XM_011543471.1 17 5966 XP_011541773.1
XM_011543473.1 17 5914 XP_011541775.1
XM_011543477.2 17 5916 XP_011541779.1
XM_017009565.1 18 6126 XP_016865054.1
XM_017009566.1 18 6016 XP_016865055.1
XM_017009567.1 16 5754 XP_016865056.1
XM_017009568.1 18 6000 XP_016865057.1
XM_017009569.1 17 5879 XP_016865058.1
XM_017009570.1 14 6699 XP_016865059.1
XM_017009571.1 17 5922 XP_016865060.1
AF536980.1 1 3507
U02882.1 14 5430 AAC13745.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv830312 Chr5:58895752 - 59086399 on Build GRCh38 Gain PDE4D

View Full Product Details