Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation ID |
Location |
CNV Subtype |
Genes |
nsv831918 |
Chr10:76703492 - 76896255 on Build GRCh38 |
Gain |
KCNMA1-AS1
KCNMA1
|
Product Details
- Assay Reference Genome
Location
Chr.10:76872732 on build GRCh38
- Cytoband
- 10q22.3
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 3778
- Gene Symbol
- KCNMA1
- Gene Name
- potassium calcium-activated channel subfamily M alpha 1
- Gene Aliases
- BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
- Location
- Chr.10:76869601-77637819 on build GRCh38
- Assay Gene Location
- Within Exon 44
Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation |
Location |
CNV Subtype |
Genes |
nsv831918 |
Chr10:76703492 - 76896255 on Build GRCh38 |
Gain |
KCNMA1-AS1
KCNMA1
|
View Full Product Details