Product Details
- Assay Reference Genome
Location
Chr.X:31120785 on build GRCh38
- Cytoband
- Xp21.2
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 1756
- Gene Symbol
- DMD
- Gene Name
- dystrophin
- Gene Aliases
- BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85
- Location
- Chr.X:31119219-33339609 on build GRCh38
- Assay Gene Location
- Within Exon 88
View Full Product Details