Species:
Human, Rhesus monkey
Mature miRNA Sequence:
CUAAUAGUAUCUACCACAAUAAA
miRBase Accession Numbers:
MIMAT0003303, MIMAT0006478
Mapped to miRBase Version:
v22.1
Megaplex RT Primer Pool:
Megaplex™ RT Primers, Human Pool B v3.0 Megaplex™ RT Primers, Human Pool B v3.0
TaqMan® Array MicroRNA Card:
TaqMan® Human MicroRNA B Cards v3.0 TaqMan® Human MicroRNA B Cards v3.0

View Full Details