Product Details
- Assay Reference Genome
Location
Chr.2:140661162 on build GRCm38
- Cytoband
- 2F3
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 71436
- Gene Symbol
- Flrt3
- Gene Name
- fibronectin leucine rich transmembrane protein 3
- Gene Aliases
- 5530600M07Rik, C430047I10Rik, mKIAA1469
- Location
- Chr.2:140658198-140671476 on build GRCm38
- Assay Gene Location
- Within Exon 3
- Entrez Gene ID
- 72899
- Gene Symbol
- Macrod2
- Gene Name
- MACRO domain containing 2
- Gene Aliases
- 1110033L15Rik, 2610107G07Rik, 2900006F19Rik
- Location
- Chr.2:140395356-142392966 on build GRCm38
- Assay Gene Location
- Within Intron 5
View Full Product Details