Product Details
- Assay Reference Genome
Location
Chr.4:85764748 on build GRCm38
- Cytoband
- 4C4
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 77739
- Gene Symbol
- Adamtsl1
- Gene Name
- ADAMTS-like 1
- Gene Aliases
- 5930437A14Rik, 6720426B09Rik, ADAMTSL-1
- Location
- Chr.4:85513234-86428390 on build GRCm38
- Assay Gene Location
Gene Symbol |
Transcript Accession |
Exon Location |
Assay Location |
Protein ID |
Adamtsl1 |
|
|
|
|
View Full Product Details