Product Details
- Assay Reference Genome
Location
Chr.11:131540052 on build GRCh38
- Cytoband
- 11q25
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 101929653
- Gene Symbol
- LOC101929653
- Gene Name
- uncharacterized LOC101929653
- Gene Aliases
- Location
- Chr.11:131533857-131540867 on build GRCh38
- Assay Gene Location
- Within Intron 2
Gene Symbol |
Transcript Accession |
Exon Location |
Assay Location |
Protein ID |
LOC101929653 |
NR_110140.1 |
|
|
|
AK128059.1 |
|
|
|
- Entrez Gene ID
- 50863
- Gene Symbol
- NTM
- Gene Name
- neurotrimin
- Gene Aliases
- HNT, IGLON2, NTRI
- Location
- Chr.11:131370326-132336822 on build GRCh38
- Assay Gene Location
- Within Intron 1
View Full Product Details