Product Details

Assay Reference Genome
Location

Chr.3:175267163 on build GRCh38
Cytoband
3q26.31
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
254827
Gene Symbol
NAALADL2
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 2
Gene Aliases
Location
Chr.3:174439669-175809510 on build GRCh38
Assay Gene Location
Within Intron 18
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NAALADL2 NM_207015.2 NP_996898.2
XM_006713560.2 XP_006713623.1
XM_011512612.2 XP_011510914.1
XM_011512613.1 XP_011510915.1
XM_011512615.2 XP_011510917.1
XM_011512616.2 XP_011510918.1
XM_011512617.2 XP_011510919.1
XM_017006070.1 XP_016861559.1
XM_017006071.1 XP_016861560.1
XM_017006072.1 XP_016861561.1
XM_017006073.1 XP_016861562.1
XM_017006074.1 XP_016861563.1
XM_017006075.1 XP_016861564.1
XM_017006076.1 XP_016861565.1
XM_017006077.1 XP_016861566.1
XM_017006078.1 XP_016861567.1
XM_017006079.1 XP_016861568.1
XM_017006080.1 XP_016861569.1
XM_017006081.1 XP_016861570.1
XM_017006082.1 XP_016861571.1
XM_017006083.1 XP_016861572.1
AJ607397.1 CAE54975.1
AK131044.1
AK308416.1
AL832144.1
AL832931.1 CAH56310.1
Entrez Gene ID
100874244
Gene Symbol
NAALADL2-AS2
Gene Name
NAALADL2 antisense RNA 2
Gene Aliases
Location
Chr.3:175235092-175271096 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NAALADL2-AS2 NR_046713.1
BQ049755.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv829800 Chr3:175252630 - 175424883 on Build GRCh38 Gain+Loss NAALADL2 MIR4789 NAALADL2-AS2
nsv592550 Chr3:175143245 - 175354813 on Build GRCh38 Loss NAALADL2 NAALADL2-AS2

View Full Product Details