Mature miRNA Sequence:
UUCCUUCUGUUGUCUGUGCAG
Amplicon Length:
81
Species:
Human
Chromosome Location:
Chr. 17: 76098019 - 76098076 [-] on Build GRCh38
Mapped to miRBase Version:
v22.1

Stem-loop Details

Stem-loop ID Stem-loop Accession # Mature miRNA ID Chromosome Location
hsa-mir-6868 MI0022715 hsa-miR-6868-3p,
hsa-miR-6868-5p
Chr. 17 - 76098019 - 76098076 [-]
on Build GRCh38

View Full Stem-loop Details