Product Details

Assay Reference Genome
Location

Chr.10:51014143 on build GRCh38
Cytoband
10q11.23
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
5592
Gene Symbol
PRKG1
Gene Name
protein kinase, cGMP-dependent, type I
Gene Aliases
AAT8, PKG, PRKG1B, PRKGR1B, cGK, cGK 1, cGK1, cGKI, cGKI-BETA, cGKI-alpha
Location
Chr.10:50991151-52298350 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PRKG1 NM_001098512.2 NP_001091982.1
AK300110.1
D45864.1 BAA08297.1
DC310312.1
DC379897.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv831873 Chr10:50950850 - 51139780 on Build GRCh38 Gain LOC102724719 PRKG1
nsv831872 Chr10:50903568 - 51058777 on Build GRCh38 Gain PRKG1

View Full Product Details