R&D Systems
Manufacturer Code:AF7449SP
Catalog # AF7449SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human t-Plasminogen Activator/tPA Ser36-Pro562 Accession # P00750 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Alteplase; Alteplase DKFZp686I03148 EC 3.4.21 EC 3.4.21.68 plasminogen activator tissue plasminogen activator tissue type PLAT reteplase tissue plasminogen activator (t-PA) tissue-type plasminogen activator TPA T-PA t-plasminogen activator tPlasminogen Activator; plasminogen/activator kringle; Reteplase; t-PA; t-plasminogen activator; Tissue-type plasminogen activator; Tissue-type plasminogen activator chain A; Tissue-type plasminogen activator chain B
Gene Aliases: PLAT; T-PA; TPA
UniProt ID: (Human) P00750
Entrez Gene ID: (Human) 5327
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.