Abnova
Manufacturer Code:H00051734B01
Catalog # 89-007-705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SEPX1 (NP_057416.1 1 a.a. ~ 94 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human SEPX1 protein.
For Research Use Only
Protein Aliases: HSPC270; Methionine-R-sulfoxide reductase B1; MGC3344; MSRB1; selenoprotein R; Selenoprotein X; selenoprotein X, 1; SELR; SELX
Gene Aliases: HSPC270; MSRB1; SELR; SELX; SepR; SEPX1
UniProt ID: (Human) Q9NZV6
Entrez Gene ID: (Human) 51734
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.