Novus Biologicals
Manufacturer Code:NBP274151IR
Catalog # NBP274151IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6C2 |
Immunogen | Recombinant protein fragment corresponding to amino acids 25-295 of human sFRP-2 (NP_003004) produced in E.coli. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FRP-2; FRP2 FRP-2secreted apoptosis related protein 1 SARP-1 SARP1sFRP-2 SDF-5 Secreted apoptosis-related protein 1 secreted frizzled-related protein 2; SARP-1; secreted apoptosis related protein 1; Secreted apoptosis-related protein 1; Secreted frizzled-related protein 2; sFRP-2; testicular tissue protein Li 170
Gene Aliases: FKSG12; FRP-2; FRP2; SARP1; SDF-5; SFRP2; UNQ361/PRO697
UniProt ID: (Human) Q96HF1
Entrez Gene ID: (Human) 6423
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.