Abnova
Manufacturer Code:H00004889B01P
Catalog # 89-016-866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | NPY5R (NP_006165.1 1 a.a. ~ 445 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human NPY5R protein.
For Research Use Only
Protein Aliases: Neuropeptide Y receptor type 5; NPY-Y5 receptor; NPY5-R; NPYR5; NPYY5-R; Y5 receptor
Gene Aliases: NPY5-R; NPY5R; NPYR5; NPYY5-R
UniProt ID: (Human) Q15761
Entrez Gene ID: (Human) 4889
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.