Abnova
Manufacturer Code:H00079877D01P
Catalog # 89-018-757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DCAKD (NP_079095.2 1 a.a. ~ 231 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human DCAKD protein.
For Research Use Only
Protein Aliases: Dephospho-CoA kinase domain-containing protein; FLJ22955
Gene Aliases: DCAKD
UniProt ID: (Human) Q8WVC6
Entrez Gene ID: (Human) 79877
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.