Invitrogen
Manufacturer Code:PA523296
Catalog # PIPA523296
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (LGKRDFVDHLDKVDPVDG (aa23-40)) of ARRB2 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARB2 ARR2 BARR2 arrestin 3 arrestin beta-2 beta-arrestin-2; arrestin 3; Arrestin beta-2; arrestin, beta 2; arrestin-3; beta arr2; beta-arrestin 2 long isoform; beta-arrestin 2 short isoform; Beta-arrestin-2; Non-visual arrestin-3
Gene Aliases: AI326910; ARB2; ARR2; ARRB2; AW122872; BARR2; BARRES
UniProt ID: (Human) P32121, (Mouse) Q91YI4, (Rat) P29067
Entrez Gene ID: (Bovine) 281638, (Human) 409, (Horse) 100072892, (Dog) 607262, (Mouse) 216869, (Rat) 25388
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.