Abnova
Manufacturer Code:H00000409M06
Catalog # 89-000-530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D2 |
Immunogen | ARRB2 (AAH07427 300 a.a. ~ 409 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ARRB2.
For Research Use Only
Protein Aliases: ARB2; ARR2; arrestin 3; Arrestin beta-2; arrestin, beta 2; BARR2; Beta-arrestin-2; DKFZp686L0365; Non-visual arrestin-3
Gene Aliases: ARB2; ARR2; ARRB2; BARR2
UniProt ID: (Human) P32121
Entrez Gene ID: (Human) 409
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.