Proteintech
Manufacturer Code:112241AP150UL
Catalog # 50-172-6981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | alpha Tubulin Fusion Protein Ag1727 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha tubulin; alpha Tubulin Alpha tubulin ubiquitous alpha tubulina tubulin TUBA1B tubulin tubulin alpha 1b; Alpha-tubulin 2; Alpha-tubulin isotype M-alpha-2; Alpha-tubulin ubiquitous; Detyrosinated tubulin alpha-1B chain; Tubulin alpha-1B chain; Tubulin alpha-2 chain; Tubulin alpha-ubiquitous chain; Tubulin K-alpha-1; tubulin, alpha 1b; tubulin, alpha 2; tubulin, alpha, ubiquitous
Gene Aliases: K-ALPHA-1; RGD1565476; TUBA1B; Tuba2
UniProt ID: (Human) P68363, (Mouse) P05213, (Rat) Q6P9V9
Entrez Gene ID: (Human) 10376, (Mouse) 22143, (Rat) 500929
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.