Invitrogen
Manufacturer Code:PA569022
Catalog # PIPA569022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human ZNF335 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MCPH10 NIF-1 NIF1 NIF2 zinc finger protein 335 NRC-interacting factor 1 zinc-finger/leucine-zipper co-transducer NIF1; NIF-1; NRC-interacting factor 1; Zinc finger protein 335; zinc-finger/leucine-zipper co-transducer NIF-1; zinc-finger/leucine-zipper co-transducer NIF1
Gene Aliases: MCPH10; Nif; NIF-1; NIF1; NIF2; Zfp335; ZNF335
UniProt ID: (Human) Q9H4Z2, (Rat) G3V893
Entrez Gene ID: (Human) 63925, (Rabbit) 100348604, (Dog) 485904, (Horse) 100071131, (Bovine) 535476, (Rat) 259270, (Guinea pig) 100721598
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.