Invitrogen
Manufacturer Code:PA521076
Catalog # PIPA521076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide near the amino terminus of human ZIP13. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LIV-1 subfamily of ZIP zinc transporter 9; LIV-1 subfamily of ZIP zinc transporter 9 LZT-Hs9 solute carrier family 39 (metal ion transporter) member 13 zinc transporter ZIP13 zrt- and Irt-like protein 13; LZT-Hs9; solute carrier family 39 (metal ion transporter), member 13; solute carrier family 39 (zinc transporter), member 13; Solute carrier family 39 member 13; Zinc transporter ZIP13; ZIP-13; Zrt- and Irt-like protein 13
Gene Aliases: LZT-Hs9; SLC39A13; ZIP13
UniProt ID: (Human) Q96H72
Entrez Gene ID: (Human) 91252
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.