OriGene
Manufacturer Code:CF801568
Catalog # 50-166-5637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H8 |
Immunogen | Full length human recombit protein of human WWC1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HBeAg-binding protein 3; HBEBP3 HBEBP36 KIBRA MEMRYQTL PPP1R168; KIBRA; Kidney and brain protein; Protein KIBRA; protein phosphatase 1, regulatory subunit 168; protein WWC1; WW domain-containing protein 1; WW, C2 and coiled-coil domain containing 1
Gene Aliases: HBEBP3; HBEBP36; KIAA0869; KIBRA; MEMRYQTL; PPP1R168; WWC1
UniProt ID: (Human) Q8IX03
Entrez Gene ID: (Human) 23286
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.