Invitrogen
Manufacturer Code:PA545620
Catalog # PIPA545620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human WWC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HBeAg-binding protein 3; KIBRA; Kidney and brain protein; Protein KIBRA; protein phosphatase 1, regulatory subunit 168; protein WWC1; WW and C2 domain containing 1; WW and C2 domain containing 1 FLJ10865 FLJ23369 HBEBP3 HBEBP36 KIAA0869 KIBRA; WW domain-containing protein 1; WW, C2 and coiled-coil domain containing 1
Gene Aliases: AA408228; AU017197; BC037006; HBEBP3; HBEBP36; KIAA0869; KIBRA; MEMRYQTL; mKIAA0869; PPP1R168; RGD1308329; WWC1
UniProt ID: (Human) Q8IX03, (Mouse) Q5SXA9
Entrez Gene ID: (Rabbit) 100358939, (Human) 23286, (Horse) 100070535, (Dog) 479297, (Rat) 303039, (Mouse) 211652, (Guinea pig) 100712753
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.