Novus Biologicals
Manufacturer Code:NBP100139
Catalog # NBP100139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SEEDHLPLQVLQSH corresponding to C-Terminus according to Uniprot Mouse NP_035836.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Osm-9-like TRP channel 2; Osm-9-like TRP channel 2 transient receptor potential cation channel subfamily V member 2 TrpV2 Vanilloid receptor-like protein 1 VRL-1MGC12549 VRL1transient receptor potential cation channel subfamily V member 2 VRLOTRPC2; OTRPC2; Transient receptor potential cation channel subfamily V member 2; transient receptor potential cation channel, subfamily V, member 2; TrpV2; Vanilloid receptor-like protein 1; VRL-1
Gene Aliases: TRPV2; VRL; VRL-1; VRL1
UniProt ID: (Human) Q9Y5S1
Entrez Gene ID: (Human) 51393
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.