Novus Biologicals
Manufacturer Code:NBP274850FR
Catalog # NBP274850FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9E11 |
Immunogen | Full-length protein expressed in 293T cell transfected with human VMAT2 expression vector. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: MGC120477 monoamine neurotransmitter transporter Monoamine transporter solute carrier family 18 (vesicular monoamine) member 2 Solute carrier family 18 member 2 SVAT SVMTMGC120478 synaptic vesicular amine transporter VAT2MGC26538 vesicle monoamine transporter type 2 vesicle monoamine/H&plus antiporter Vesicular amine transporter 2 VMAT2; monoamine neurotransmitter transporter; Monoamine transporter; solute carrier family 18 (vesicular monoamine transporter), member 2; solute carrier family 18 (vesicular monoamine), member 2; Solute carrier family 18 member 2; Synaptic vesicular amine transporter; VAT2; vesicle monoamine transporter type 2; vesicle monoamine/H+ antiporter; Vesicular amine transporter 2
Gene Aliases: SLC18A2; SVAT; SVMT; VAT2; VMAT2
UniProt ID: (Human) Q05940
Entrez Gene ID: (Human) 6571
Molecular Function: amino acid transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.