Invitrogen
Manufacturer Code:PA5102661
Catalog # PIPA5102661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC18A2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110037L13Rik 9330105E13 MGC120477 MGC120478 MGC26538 MNAT monoamine neurotransmitter transporter monoamine transporter SLC18A2 solute carrier family 18 (vesicular monoamine transporter) member 2 solute carrier family 18 (vesicular monoamine) member 2 Solute carrier family 18 A2 (vesicular monoamine transporter 2) solute carrier family 18 member 2 solute carrier family 18 member A2 solute carrier family 18 member 2 SVAT SVMT Synaptic vesicular amine transporter VAT2 vesicle monoamine transporter type 2 vesicle monoamine/H+ antiporter vesicular amine transporter 2 vesicular monoamine transporter 2 VMAT2 VMAT-2; monoamine neurotransmitter transporter; Monoamine transporter; solute carrier family 18 (vesicular monoamine transporter), member 2; solute carrier family 18 (vesicular monoamine), member 2; Solute carrier family 18 A2 (vesicular monoamine transporter 2); Solute carrier family 18 member 2; solute carrier family 18, member 2; Synaptic vesicular amine transporter; VAT2; vesicle monoamine transporter type 2; vesicle monoamine/H+ antiporter; Vesicular amine transporter 2; vesicular monoamine transporter 2
Gene Aliases: 1110037L13Rik; 9330105E13; MNAT; SLC18A2; SVAT; SVMT; VAT2; VMAT-2; VMAT2
UniProt ID: (Human) Q05940, (Mouse) Q8BRU6, (Rat) Q01827
Entrez Gene ID: (Human) 6571, (Mouse) 214084, (Rat) 25549
Molecular Function: amino acid transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.