Novus Biologicals
Manufacturer Code:NBP274863APCCY7
Catalog # N274863APC7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B8 |
Immunogen | Full length recombinant protein of human V-type proton ATPase subunit F (NP_004222) produced in E.coli. |
Conjugate | APC-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: adenosinetriphosphatase 14k chain; ATP6S14adenosinetriphosphatase 14k chain ATPase H&plus transporting lysosomal 14kDa V1 subunit F H(&plus)-transporting two-sector ATPase 14kD subunit MGC117321 MGC126037 MGC126038 vacuolar ATP synthase subunit F vacuolar proton pump F subunit Vacuolar proton pump subunit F VATFATPase vacuolar 14 kD V-ATPase 14 kDa subunit V-ATPase F subunit V-ATPase subunit F Vma7 V-type proton ATPase subunit F; ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F; ATPase, vacuolar, 14 kD; H(+)-transporting two-sector ATPase, 14kD subunit; V-ATPase 14 kDa subunit; V-ATPase F subunit; V-ATPase subunit F; V-type proton ATPase subunit F; vacuolar ATP synthase subunit F; vacuolar proton pump F subunit; Vacuolar proton pump subunit F
Gene Aliases: ATP6S14; ATP6V1F; VATF; Vma7
UniProt ID: (Human) Q16864
Entrez Gene ID: (Human) 9296
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.