Novus Biologicals
Manufacturer Code:NBP297933FR
Catalog # NB049591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human USPL1 fragment. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA121O19.1 C13orf22 chromosome 13 open reading frame 22 D13S106 D13S106E DKFZp781K2286 FLJ32952 highly charged protein RP11-121O19.1 ubiquitin specific peptidase like 1 ubiquitin-specific peptidase-like protein 1; highly charged protein; SUMO-specific isopeptidase USPL1; Ubiquitin-specific peptidase-like protein 1; USP-like 1
Gene Aliases: bA121O19.1; C13orf22; D13S106; D13S106E; USPL1
UniProt ID: (Human) Q5W0Q7
Entrez Gene ID: (Human) 10208
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.