Invitrogen
Manufacturer Code:PA543134
Catalog # PIPA543134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human UMPS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mutant uridine monophosphate synthase; ODC; OMPdecase; OPRT; OPRTase; orotate phosphoribosyl transferase and orotidine-5'-decarboxylase; Orotate phosphoribosyltransferase; Orotidine 5'-phosphate decarboxylase; UMP synthase; Uridine 5'-monophosphate synthase; uridine 5'-monophosphate synthase-like protein; uridine monophophate synthase; uridine monophosphate synthase; uridine monophosphate synthetase (orotate phosphoribosyl transferase and orotidine-5'-decarboxylase); Uridine monophosphate synthetase OPRT
Gene Aliases: 1700095D23Rik; AA408257; AL033308; BB164745; OK/SW-cl.21; OPRT; UMPS
UniProt ID: (Human) P11172, (Bovine) P31754, (Mouse) P13439
Entrez Gene ID: (Horse) 100070386, (Pig) 100516340, (Rabbit) 100353794, (Human) 7372, (Dog) 478593, (Bovine) 281568, (Mouse) 22247, (Guinea pig) 100726089, (Rat) 288051
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.