Novus Biologicals
Manufacturer Code:NBP170740
Catalog # NBP170740
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to C4ORF20 The peptide sequence was selected from the middle region of C4ORF20. Peptide sequence YHHYMQDRIDDNGWGCAYRSLQTICSWFKHQGYTERSIPTHREIQQALVD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ11200 UFM1-specific peptidase 2 ufm1-specific protease 2; UFM1-specific peptidase 2; Ufm1-specific protease 2; UfSP2
Gene Aliases: BHD; C4orf20; UFSP2
UniProt ID: (Human) Q9NUQ7
Entrez Gene ID: (Human) 55325
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.