Invitrogen
Manufacturer Code:MA535505
Catalog # PIMA535505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0996 |
Immunogen | A synthesized peptide derived from human UBE2I |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 5830467E05Rik C358B7.1 F830028O17Rik mUBC9 OTTHUMP00000198131 p18 RING-type E3 SUMO transferase UBC9 SUMO-conjugating enzyme UBC9 SUMO-protein ligase Ubc9 UbcE2A Ubce2i Ubce9 UBE2I Ubiquitin carrier protein 9 Ubiquitin carrier protein I Ubiquitin-conjugating enzyme E2 I Ubiquitin-conjugating enzyme UbcE2A Ubiquitin-protein ligase I; mUBC9; p18; RING-type E3 SUMO transferase UBC9; SUMO-1-protein ligase; SUMO-conjugating enzyme UBC9; SUMO-protein ligase; Ubiquitin carrier protein 9; Ubiquitin carrier protein I; ubiquitin conjugating enzyme 9; ubiquitin conjugating enzyme E2I; Ubiquitin conjugating enzyme E2I (homologous to yeast UBC9); Ubiquitin-conjugating enzyme E2 I; ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9); ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast); Ubiquitin-conjugating enzyme UbcE2A; ubiquitin-like protein SUMO-1 conjugating enzyme; ubiquitin-protein ligase E2I; Ubiquitin-protein ligase I
Gene Aliases: 5830467E05Rik; C358B7.1; F830028O17Rik; P18; UBC9; UbcE2A; Ubce2i; UBCE9; UBE2I
UniProt ID: (Human) P63279, (Rat) P63281, (Mouse) P63280
Entrez Gene ID: (Human) 7329, (Rat) 25573, (Mouse) 22196
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.