Invitrogen
Manufacturer Code:PA519609
Catalog # PIPA519609
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 100 to the C-terminus of Human UBE2I / UBC9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C358B7.1 P18 UBC9 SUMO-1-protein ligase SUMO-conjugating enzyme UBC9 SUMO-protein ligase ubiquitin carrier protein 9 ubiquitin carrier protein I ubiquitin conjugating enzyme 9 ubiquitin-conjugating enzyme E2 I ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9) ubiquitin-conjugating enzyme UbcE2A ubiquitin-like protein SUMO-1 conjugating enzyme ubiquitin-protein ligase E2I ubiquitin-protein ligase I; mUBC9; p18; RING-type E3 SUMO transferase UBC9; SUMO-1-protein ligase; SUMO-conjugating enzyme UBC9; SUMO-protein ligase; Ubiquitin carrier protein 9; Ubiquitin carrier protein I; ubiquitin conjugating enzyme 9; ubiquitin conjugating enzyme E2I; Ubiquitin conjugating enzyme E2I (homologous to yeast UBC9); Ubiquitin-conjugating enzyme E2 I; ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9); ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast); Ubiquitin-conjugating enzyme UbcE2A; ubiquitin-like protein SUMO-1 conjugating enzyme; ubiquitin-protein ligase E2I; Ubiquitin-protein ligase I
Gene Aliases: 5830467E05Rik; C358B7.1; F830028O17Rik; P18; UBC9; UbcE2A; Ubce2i; UBCE9; UBE2I
UniProt ID: (Human) P63279, (Rat) P63281, (Mouse) P63280
Entrez Gene ID: (Human) 7329, (Rat) 25573, (Mouse) 22196
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.