Novus Biologicals
Manufacturer Code:NBP250402F
Catalog # NBP250402F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | F24 |
Immunogen | Purified chicken breast muscle |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: beta TnTF; Beta-TnTF; Beta-TnTF fast skeletal muscle TnTf troponin T type 3 (skeletal fast) troponin T3 skeletal fast troponin-T3 skeletal fast; Fast skeletal muscle troponin T; fTnT; TnTf; troponin T type 3 (skeletal, fast); Troponin T, fast skeletal muscle; troponin-T3, skeletal, fast
Gene Aliases: TNNT3; TNTF
UniProt ID: (Human) P45378
Entrez Gene ID: (Human) 7140
Molecular Function: actin binding motor protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.