R&D Systems
Manufacturer Code:IC7428T100UG
Catalog # IC7428T100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 489804 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse Thioredoxin Reductase 1/TRXR1 Met1-Ile497 Accession # Q16881 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: EC 1.8.1 EC 1.8.1.9 Gene associated with retinoic and IFN-induced mortality 12 protein Gene associated with retinoic and interferon-induced mortality 12 protein GRIM12 GRIM-12 GRIM-12TR1 KDRF KM-102-derived reductase-like factor MGC9145 oxidoreductase thioredoxin reductase 1 thioredoxin reductase 1 cytoplasmic thioredoxin reductase GRIM-12 Thioredoxin reductase TR1 TR TR1 Trxr1 TRXRD1 TXNR TXNRD1 TXNRTRXR1; gene associated with retinoic and IFN-induced mortality 12 protein; Gene associated with retinoic and interferon-induced mortality 12 protein; GRIM-12; KM-102-derived reductase-like factor; oxidoreductase; testis tissue sperm-binding protein Li 46a; Thioredoxin reductase 1, cytoplasmic; thioredoxin reductase GRIM-12; Thioredoxin reductase TR1; TR
Gene Aliases: GRIM-12; GRIM12; KDRF; TR; TR1; TRXR1; TXNR; TXNRD1
UniProt ID: (Human) Q6VBP2
Entrez Gene ID: (Human) 7296
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.