Abnova
Manufacturer Code:MAB0686
Catalog # 89-108-042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7B2 |
Immunogen | Recombinant protein corresponding to amino acids 37-524 of human TXNRD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant TXNRD2.
For Research Use Only
Protein Aliases: Selenoprotein Z; SELZ; Thioredoxin reductase 2, mitochondrial; thioredoxin reductase 3; thioredoxin reductase beta; Thioredoxin reductase TR3; TR; TR-BETA; TR3; TRXR2
Gene Aliases: KIAA1652; SELZ; TR; TR-BETA; TR3; TRXR2; TXNRD2
UniProt ID: (Human) Q9NNW7
Entrez Gene ID: (Human) 10587
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.