Invitrogen
Manufacturer Code:PA585593
Catalog # PIPA585593
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human Transcription Termination Factor 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RNA polymerase I termination factor; Transcription termination factor 1; Transcription termination factor I; transcription termination factor, RNA polymerase I; TTF-1; TTF-I; TTF-I Antibody transcription termination factor RNA polymerase I Antibody TTF-1 Antibody
Gene Aliases: TTF-1; TTF-I; TTF1
UniProt ID: (Human) Q15361
Entrez Gene ID: (Human) 7270
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.