Invitrogen
Manufacturer Code:PA5111808
Catalog # PIPA5111808
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunomicroscopy (IM) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Guinea pig |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 413-428 of rat TRPV2 (1stextracellular loop) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Guinea Pig Polyclonal Antibody
For Research Use Only
Protein Aliases: Grc Growth factor-regulated calcium channel Osm-9-like TRP channel 2 OTRPC2 RP23-234K24.7 Sac2b Stretch-activated channel 2B Transient receptor potential cation channel subfamily V member 2 Trpv2 Vanilloid receptor-like protein 1 VRL Vrl1 VRL-1; Osm-9-like TRP channel 2; OTRPC2; Stretch-activated channel 2B; Transient receptor potential cation channel subfamily V member 2; TrpV2; Vanilloid receptor-like protein 1; VRL-1
Gene Aliases: Sac2b; Trpv2; Vrl1
UniProt ID: (Rat) Q9WUD2
Entrez Gene ID: (Rat) 29465
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.