Invitrogen
Manufacturer Code:OSR00077W
Catalog # PIOSR00077W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 4th cytoplasmic loop of rat TRPM8 conjugated to an immunogenic carrier protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cold menthol receptor 1; cold/menthol receptor 1; Transient receptor potential cation channel subfamily M member 8; Transient receptor potential cation channel subfamily M member 8 Cold menthol receptor 1 Trpm8 Trpm8 transient receptor potential cation channel s
Gene Aliases: CMR1; Trpm8
UniProt ID: (Rat) Q8R455
Entrez Gene ID: (Rat) 171384
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.