Novus Biologicals
Manufacturer Code:NBP274596V
Catalog # NBP274596V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8C8 |
Immunogen | Full length recombinant protein of human TRIB1 (NP_079471) produced in HEK293T cells. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: C8FWG-protein-coupled receptor induced protein GIG-2 GIG2G-protein-coupled receptor-induced protein 2 G-protein-coupled receptor-induced gene 2 protein SKIP1 TRB-1 TRB1phosphoprotein regulated by mitogenic pathways tribbles homolog 1 tribbles homolog 1 (Drosophila) tribbles-like protein 1; G-protein-coupled receptor induced protein; G-protein-coupled receptor-induced gene 2 protein; G-protein-coupled receptor-induced protein 2; GIG-2; phosphoprotein regulated by mitogenic pathways; SKIP1; TRB-1; TRIB1; Tribbles homolog 1; tribbles-like protein 1
Gene Aliases: C8FW; GIG-2; GIG2; SKIP1; TRB-1; TRB1; TRIB1
UniProt ID: (Human) Q96RU8
Entrez Gene ID: (Human) 10221
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.