Invitrogen
Manufacturer Code:PA544630
Catalog # PIPA544630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TRAPPC4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hematopoietic stem/progenitor cell protein 172; Synbindin; syndecan binding protein 2; Trafficking protein particle complex subunit 4; TRS23 homolog
Gene Aliases: 1500017G03Rik; AI303617; CGI-104; HSPC172; PTD009; Sbd; SBDN; Sdcbp2; SYNBINDIN; TRAPPC4; TRS23; zgc:73260
UniProt ID: (Human) Q9Y296, (Bovine) Q2TBL9, (Rat) Q69BT7, (Mouse) Q9ES56
Entrez Gene ID: (Human) 51399, (Bovine) 785345, (Dog) 479411, (Rabbit) 100357700, (Horse) 100071306, (Rat) 367073, (Guinea pig) 100722363, (Mouse) 60409, (Zebrafish) 393737
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.