Invitrogen
Manufacturer Code:PA543538
Catalog # PIPA543538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TOR2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Prosalusin; Salusin-alpha; Salusin-beta; Torsin family 2 member A; torsin family 2, member A; Torsin-2A; Torsin-related protein 1
Gene Aliases: HEMBA1005096; Prosalusin; PSEC0218; TOR2A; TORP1; Torsin-2A; UNQ6408/PRO21181
UniProt ID: (Human) Q5JU69, (Bovine) A4FUH1, (Mouse) Q8R1J9, (Rat) Q6AYR4
Entrez Gene ID: (Human) 27433, (Dog) 609191, (Horse) 100070462, (Bovine) 534311, (Mouse) 30933, (Rat) 362112, (Guinea pig) 100726394
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.