Invitrogen
Manufacturer Code:PA585675
Catalog # PIPA585675
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human Troponin T fast skeletal. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta TnTF; Beta-TnTF; Fast skeletal muscle troponin T; fast skeletal muscle troponin T3; fast skeletal troponin T beta-1 isoform; fTnT; skeletal muscle fast-twitch TnT; skeletal troponin T alpha-1 isoform; TnTf; troponin T type 3 (skeletal fast) Antibody TNTF Antibody; troponin T type 3 (skeletal, fast); Troponin T, fast skeletal muscle; troponin T3, skeletal, fast; troponin-T3, skeletal, fast
Gene Aliases: fTnT; TNNT3; Tnt; TNTF
UniProt ID: (Human) P45378, (Rat) P09739, (Mouse) O35585
Entrez Gene ID: (Human) 7140, (Rat) 24838, (Mouse) 21957
Molecular Function: actin binding motor protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.